Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 4 | 1229 | ||
NM_005916.4 | NP_005907.3 | 5 | 1154 | |||
NM_182776.2 | NP_877577.1 | 4 | 1120 | |||
XM_005250348.3 | XP_005250405.1 | 5 | 808 | |||
XM_017012217.1 | XP_016867706.1 | 5 | 685 | |||
GenBank mRNA | AK055379.1 | - | 4 | 1097 | ||
AK096959.1 | - | 6 | 518 | |||
AK226175.1 | - | 4 | 659 | |||
AK293172.1 | - | 5 | 556 | |||
AK314575.1 | - | 5 | 621 | |||
BC009398.2 | - | 5 | 572 | 4134871 | ||
BC013375.2 | - | 5 | 590 | 3946342 | ||
D55716.1 | - | 5 | 624 | |||
D86748.1 | - | 5 | 594 | |||
KU178172.1 | - | 5 | 508 | |||
KU178173.1 | - | 4 | 583 | |||
X74796.1 | - | 2 | 204 |