Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 10 | 1996 | ||
NM_005916.4 | NP_005907.3 | 11 | 1921 | |||
NM_182776.2 | NP_877577.1 | 10 | 1887 | |||
XM_005250348.3 | XP_005250405.1 | 11 | 1575 | |||
XM_017012217.1 | XP_016867706.1 | 11 | 1452 | |||
GenBank mRNA | AK055379.1 | - | 10 | 1864 | ||
AK096959.1 | - | 12 | 1285 | |||
AK226175.1 | - | 10 | 1426 | |||
AK293172.1 | - | 11 | 1323 | |||
AK314575.1 | - | 11 | 1388 | |||
AY007130.1 | - | 3 | 218 | |||
BC013375.2 | - | 11 | 1357 | 3946342 | ||
D55716.1 | - | 11 | 1391 | |||
D86748.1 | - | 11 | 1361 | |||
KU178172.1 | - | 11 | 1275 | |||
KU178173.1 | - | 10 | 1350 | |||
X74796.1 | - | 8 | 971 |