Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012328.2 | NP_036460.1 | 2 | 525 | ||
GenBank mRNA | AB026908.1 | - | 2 | 358 | ||
AF083247.1 | - | 2 | 359 | |||
AF093824.1 | - | 2 | 325 | |||
AK092204.1 | - | 3 | 383 | |||
AL080081.1 | - | 2 | 405 | |||
AY359045.1 | - | 1 | 179 | |||
BC028912.1 | - | 2 | 339 | 4943933 | ||
CR533475.1 | - | 1 | 156 |