Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204171.1 | NP_001191100.1 | 9 | 878 | ||
NM_001278516.1 | NP_001265445.1 | 9 | 960 | |||
NM_001278517.1 | NP_001265446.1 | 7 | 734 | |||
NM_001278518.1 | NP_001265447.1 | 6 | 549 | |||
NM_001278519.1 | NP_001265448.1 | 4 | 359 | |||
NM_002393.4 | NP_002384.2 | 10 | 1028 | |||
XM_017001311.1 | XP_016856800.1 | 10 | 1027 | |||
XM_017001312.1 | XP_016856801.1 | 10 | 1012 | |||
XM_017001313.1 | XP_016856802.1 | 9 | 877 | |||
GenBank mRNA | AB209383.1 | - | 1 | 2592 | ||
AF007111.1 | - | 10 | 978 | |||
AK223228.1 | - | 10 | 991 | |||
AK304698.1 | - | 8 | 784 | |||
AK310607.1 | - | 10 | 1004 | |||
AK310903.1 | - | 9 | 933 | |||
AY949028.1 | - | 8 | 794 | |||
BC067299.1 | - | 10 | 1018 | 30390159 | ||
BC105106.1 | - | 9 | 771 | 8144109 | ||
BC143431.1 | - | 9 | 853 | |||
BC143432.1 | - | 10 | 950 | |||
BX640923.1 | - | 10 | 973 | |||
CR749470.1 | - | 9 | 955 |