Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168335.1 | NP_001161807.1 | 6, 7 | 885 | ||
NM_002396.4 | NP_002387.1 | 6, 7 | 885 | |||
GenBank mRNA | AK313391.1 | - | 6, 7 | 781 | ||
BC000147.1 | - | 6, 7 | 827 | 2900677 | ||
M55905.1 | - | 6, 7 | 715 |