Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002398.2 | NP_002389.1 | 10, 11 | 1488 | ||
XM_005264321.2 | XP_005264378.1 | 11, 12 | 1811 | |||
XM_005264322.2 | XP_005264379.1 | 10, 11 | 1763 | |||
XM_005264323.2 | XP_005264380.1 | 11, 12 | 1811 | |||
XM_005264324.4 | XP_005264381.1 | 9, 10 | 884 | |||
XM_005264325.4 | XP_005264382.1 | 9, 10 | 2494 | |||
XM_017004144.1 | XP_016859633.1 | 10, 11 | 1669 | |||
XM_017004145.1 | XP_016859634.1 | 9, 10 | 1621 | |||
XM_017004146.1 | XP_016859635.1 | 9, 10 | 1621 | |||
GenBank mRNA | AK098174.1 | - | 8, 9 | 865 | ||
AK297109.1 | - | 9, 10 | 1143 | |||
AK297817.1 | - | 8, 9 | 803 | |||
AK314949.1 | - | 10, 11 | 1238 | |||
AL832770.1 | - | 10, 11 | 1501 | |||
BC043503.1 | - | 10, 11 | 1483 | 5531644 | ||
U85707.1 | - | 10, 11 | 1096 |