Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005923.3 | NP_005914.1 | 7 | 1518 | ||
XM_011535839.2 | XP_011534141.2 | 8 | 1842 | |||
XM_017010870.1 | XP_016866359.1 | 8 | 1841 | |||
XM_017010871.1 | XP_016866360.1 | 8 | 1841 | |||
XM_017010872.1 | XP_016866361.1 | 8 | 1840 | |||
XM_017010873.1 | XP_016866362.1 | 8 | 1839 | |||
XM_017010874.1 | XP_016866363.1 | 8 | 1841 | |||
XM_017010875.1 | XP_016866364.1 | 8 | 1840 | |||
XM_017010876.1 | XP_016866365.1 | 6 | 849 | |||
XM_017010877.1 | XP_016866366.1 | 6 | 1086 | |||
GenBank mRNA | AB209093.1 | - | 7 | 1622 | ||
AK293953.1 | - | 7 | 778 | |||
AK310942.1 | - | 8 | 1297 | |||
BC054503.2 | - | 7 | 1465 | 6143181 | ||
BC088829.1 | - | 7 | 1519 | 6007002 | ||
D84476.2 | - | 7 | 1424 | |||
U67156.1 | - | 7 | 1517 |