Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005923.3 | NP_005914.1 | 4 | 994 | ||
XM_011535839.2 | XP_011534141.2 | 5 | 1318 | |||
XM_017010870.1 | XP_016866359.1 | 5 | 1317 | |||
XM_017010871.1 | XP_016866360.1 | 5 | 1317 | |||
XM_017010872.1 | XP_016866361.1 | 5 | 1316 | |||
XM_017010873.1 | XP_016866362.1 | 5 | 1315 | |||
XM_017010874.1 | XP_016866363.1 | 5 | 1317 | |||
XM_017010875.1 | XP_016866364.1 | 5 | 1316 | |||
XM_017010876.1 | XP_016866365.1 | 3 | 325 | |||
XM_017010877.1 | XP_016866366.1 | 3 | 562 | |||
GenBank mRNA | AB209093.1 | - | 4 | 1098 | ||
AK293953.1 | - | 4 | 254 | |||
AK310942.1 | - | 5 | 773 | |||
BC054503.2 | - | 4 | 941 | 6143181 | ||
BC088829.1 | - | 4 | 995 | 6007002 | ||
D84476.2 | - | 4 | 900 | |||
U67156.1 | - | 4 | 993 |