Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098270.1 | NP_001091740.1 | 1 | 1474 | ||
NM_002409.4 | NP_002400.3 | 2 | 1589 | |||
XM_005261608.4 | XP_005261665.1 | 3 | 1624 | |||
XM_005261609.2 | XP_005261666.1 | 2 | 1730 | |||
XM_006724252.2 | XP_006724315.1 | 2 | 1434 | |||
XM_011530184.2 | XP_011528486.1 | 2 | 1462 | |||
XM_011530185.2 | XP_011528487.1 | 2 | 2813 | |||
XM_011530186.2 | XP_011528488.1 | 2 | 4459 | |||
XM_011530187.2 | XP_011528489.1 | 2 | 2046 | |||
XM_011530188.2 | XP_011528490.1 | 2 | 1458 | |||
XM_011530189.2 | XP_011528491.1 | 2 | 1379 | |||
XM_011530190.2 | XP_011528492.1 | 2 | 4988 | |||
XM_011530191.2 | XP_011528493.1 | 2 | 1483 | |||
XM_011530192.2 | XP_011528494.1 | 2 | 1375 | |||
XM_011530194.2 | XP_011528496.1 | 2 | 1375 | |||
XM_017028802.1 | XP_016884291.1 | 2 | 3307 | |||
GenBank mRNA | AK313664.1 | - | 1 | 1442 | ||
BC075025.2 | - | 2 | 1376 | 30915623 | ||
BC075026.2 | - | 2 | 1376 | 30915412 | ||
BC113383.1 | - | 2 | 1376 | 8322435 | ||
BC113718.1 | - | 2 | 1376 | 8322770 | ||
CR456519.1 | - | 1 | 1374 | |||
D13789.1 | - | 1 | 1352 |