Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002412.4 | NP_002403.2 | 5 | 627 | ||
XM_005252682.2 | XP_005252739.1 | 5 | 664 | |||
XM_017016275.1 | XP_016871764.1 | 4 | 450 | |||
GenBank mRNA | AK293593.1 | - | 5 | 597 | ||
BC000824.1 | - | 5 | 555 | 3454207 | ||
BI520029.1 | - | 5 | 609 | |||
BT006714.1 | - | 4 | 502 | |||
M29971.1 | - | 5 | 598 | |||
M31767.1 | - | 4 | 524 | |||
M60761.1 | - | 4 | 517 | |||
X54228.1 | - | 4 | 542 |