Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204366.1 | NP_001191295.1 | 5 | 681 | ||
NM_001204367.1 | NP_001191296.1 | 4 | 581 | |||
NM_001204368.1 | NP_001191297.1 | 4 | 610 | |||
NM_002413.4 | NP_002404.1 | 5 | 681 | |||
XM_006714221.2 | XP_006714284.1 | 4 | 368 | |||
XM_011531972.2 | XP_011530274.1 | 4 | 347 | |||
XM_017008211.1 | XP_016863700.1 | 5 | 693 | |||
XM_017008212.1 | XP_016863701.1 | 5 | 693 | |||
XM_017008213.1 | XP_016863702.1 | 5 | 693 | |||
XM_017008214.1 | XP_016863703.1 | 4 | 368 | |||
GenBank mRNA | AK130948.1 | - | 5 | 630 | ||
AK307231.1 | - | 4 | 416 | |||
BC025416.1 | - | 5 | 595 | 3937750 | ||
DB257575.1 | - | 5 | 445 | |||
U77604.1 | - | 5 | 440 |