Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122898.2 | NP_001116370.1 | 4 | 404 | ||
NM_001321367.1 | NP_001308296.1 | 5 | 452 | |||
NM_001321368.1 | NP_001308297.1 | 5 | 452 | |||
NM_001321369.1 | NP_001308298.1 | 4 | 404 | |||
NM_001321370.1 | NP_001308299.1 | 5 | 452 | |||
NM_002414.4 | NP_002405.1 | 5 | 452 | |||
NR_135623.1 | 5 | 452 | ||||
GenBank mRNA | AK313617.1 | - | 5 | 319 | ||
BC002584.2 | - | 5 | 330 | 3161673 | ||
BC003147.1 | - | 5 | 288 | 2906143 | ||
BC010109.2 | - | 5 | 398 | 3606974 | ||
BC021620.1 | - | 5 | 288 | 2957883 | ||
BC024310.1 | - | 5 | 391 | 4094268 | ||
BI757712.1 | - | 5 | 300 | |||
CN299062.1 | - | 4 | 210 | |||
CR450286.1 | - | 5 | 224 | |||
CX869233.1 | - | 5 | 452 | |||
DT216927.1 | - | 4 | 360 | |||
M16279.1 | - | 5 | 401 | |||
U82164.1 | - | 5 | 224 | |||
X16996.1 | - | 5 | 347 |