Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130156.2 | NP_001123628.1 | 4, 5 | 597 | ||
NM_001130157.2 | NP_001123629.1 | 5, 6 | 670 | |||
NM_001195432.1 | NP_001182361.1 | 6, 7 | 872 | |||
NM_001195434.1 | NP_001182363.1 | 6, 7 | 715 | |||
NM_022443.4 | NP_071888.1 | 4, 5 | 553 | |||
XM_005247479.4 | XP_005247536.1 | 5, 6 | 600 | |||
XM_005247480.2 | XP_005247537.1 | 5, 6 | 584 | |||
XM_006713645.2 | XP_006713708.1 | 3, 4 | 391 | |||
XM_011512852.2 | XP_011511154.1 | 6, 7 | 673 | |||
XM_011512853.2 | XP_011511155.1 | 5, 6 | 620 | |||
XM_011512854.2 | XP_011511156.1 | 3, 4 | 389 | |||
GenBank mRNA | AK056948.1 | - | 6, 7 | 685 | ||
AK096889.1 | - | 6, 7 | 781 | |||
AK297488.1 | - | 4, 5 | 539 | |||
AK315402.1 | - | 4, 5 | 523 | |||
AY848701.1 | - | 5, 6 | 639 | |||
AY848702.1 | - | 4, 5 | 566 | |||
BC007045.1 | - | 4, 5 | 522 | 3996145 | ||
BX647666.1 | - | 5, 6 | 608 | |||
KU178186.1 | - | 4, 5 | 416 | |||
KU178187.1 | - | 5, 6 | 461 | |||
KU178188.1 | - | 3, 4 | 268 | |||
L49054.1 | - | 4, 5 | 524 |