Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130156.2 | NP_001123628.1 | 5 | 623 | ||
NM_001130157.2 | NP_001123629.1 | 6 | 696 | |||
NM_001195432.1 | NP_001182361.1 | 7 | 898 | |||
NM_001195433.1 | NP_001182362.1 | 5 | 567 | |||
NM_001195434.1 | NP_001182363.1 | 7 | 741 | |||
NM_022443.4 | NP_071888.1 | 5 | 579 | |||
XM_005247479.4 | XP_005247536.1 | 6 | 626 | |||
XM_005247480.2 | XP_005247537.1 | 6 | 610 | |||
XM_006713645.2 | XP_006713708.1 | 4 | 417 | |||
XM_011512852.2 | XP_011511154.1 | 7 | 699 | |||
XM_011512853.2 | XP_011511155.1 | 6 | 646 | |||
XM_011512854.2 | XP_011511156.1 | 4 | 415 | |||
GenBank mRNA | AK056948.1 | - | 7 | 711 | ||
AK096889.1 | - | 7 | 807 | |||
AK297488.1 | - | 5 | 565 | |||
AK315402.1 | - | 5 | 549 | |||
AY848700.1 | - | 5 | 536 | |||
AY848701.1 | - | 6 | 665 | |||
AY848702.1 | - | 5 | 592 | |||
BC007045.1 | - | 5 | 548 | 3996145 | ||
BX647666.1 | - | 6 | 634 | |||
KU178186.1 | - | 5 | 442 | |||
KU178187.1 | - | 6 | 487 | |||
KU178188.1 | - | 4 | 294 | |||
L49054.1 | - | 5 | 550 |