Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166693.2 | NP_001160165.1 | 12 | 2649 | ||
NM_001313959.1 | NP_001300888.1 | 11 | 2773 | |||
NM_001313960.1 | NP_001300889.1 | 9 | 1752 | |||
NM_005935.3 | NP_005926.1 | 11 | 2760 | |||
XM_005263007.3 | XP_005263064.1 | 12 | 2740 | |||
XM_005263009.3 | XP_005263066.1 | 12 | 2641 | |||
XM_005263011.3 | XP_005263068.1 | 11 | 2435 | |||
XM_005263013.3 | XP_005263070.1 | 12 | 2644 | |||
XM_011531973.2 | XP_011530275.1 | 12 | 2562 | |||
XM_017008215.1 | XP_016863704.1 | 10 | 1606 | |||
XM_017008216.1 | XP_016863705.1 | 8 | 1993 | |||
GenBank mRNA | AK296243.1 | - | 9 | 1471 | ||
AK300364.1 | - | 12 | 2640 | |||
BC114930.1 | - | 11 | 2763 | 40074243 | ||
L13773.1 | - | 11 | 2773 | |||
L25050.1 | - | 11 | 2365 |