Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145938.1 | NP_001139410.1 | 5 | 739 | ||
NM_002421.3 | NP_002412.1 | 5 | 891 | |||
GenBank mRNA | AK024818.1 | - | 5 | 814 | ||
AK098450.1 | - | 6 | 746 | |||
AK130687.1 | - | 5 | 814 | |||
AK223034.1 | - | 4 | 517 | |||
AK223035.1 | - | 5 | 772 | |||
AK223036.1 | - | 3 | 355 | |||
AK223281.1 | - | 5 | 814 | |||
AK225495.1 | - | 4 | 582 | |||
AK297723.1 | - | 5 | 662 | |||
AK301340.1 | - | 5 | 604 | |||
BC013118.1 | - | 1 | 39 | |||
BC013875.2 | - | 5 | 790 | 3834572 | ||
BT006874.1 | - | 5 | 748 | |||
BT020147.1 | - | 5 | 748 | |||
M13509.1 | - | 5 | 816 | |||
X05231.1 | - | 5 | 814 | |||
X54925.1 | - | 5 | 816 |