Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278594.1 | NP_001265523.1 | 4 | 420 | ||
NM_005589.3 | NP_005580.1 | 4 | 420 | |||
XM_017021331.1 | XP_016876820.1 | 2 | 199 | |||
GenBank mRNA | AF038011.1 | - | 4 | 300 | ||
AF148505.1 | - | 4 | 363 | |||
AF159889.1 | - | 4 | 321 | |||
AJ249994.1 | - | 4 | 351 | |||
AK222687.1 | - | 4 | 358 | |||
AK223244.1 | - | 4 | 332 | |||
AK311478.1 | - | 4 | 376 | |||
AK312389.1 | - | 4 | 363 | |||
BC004909.1 | - | 4 | 352 | 3530715 | ||
BC032371.1 | - | 4 | 420 | 5163797 |