Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004531.4 | NP_004522.1 | 3 | 736 | ||
NM_176806.3 | NP_789776.1 | 3 | 275 | |||
GenBank mRNA | AF091871.1 | - | 3 | 248 | ||
AF117815.1 | - | 3 | 259 | |||
AF155659.1 | - | 3 | 461 | |||
AK126211.1 | - | 1 | 968 | |||
AK312887.1 | - | 3 | 261 | |||
BC029287.1 | - | 3 | 238 | |||
BC039720.2 | - | 2 | 216 | |||
BC046097.1 | - | 3 | 422 | 5740329 | ||
BC095417.1 | - | 3 | 276 | |||
CD514049.1 | - | 3 | 271 | |||
CR457172.1 | - | 1 | 45 |