Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004196.3 | NP_001004196.2 | 5 | 808 | ||
NM_001318826.1 | NP_001305755.1 | 4 | 728 | |||
NM_001318828.1 | NP_001305757.1 | 3 | 651 | |||
NM_001318830.1 | NP_001305759.1 | 3 | 646 | |||
NM_005944.6 | NP_005935.4 | 4 | 733 | |||
XM_005247481.4 | XP_005247538.1 | 5 | 982 | |||
XM_005247482.2 | XP_005247539.1 | 4 | 612 | |||
XM_005247485.2 | XP_005247542.1 | 4 | 923 | |||
XM_005247486.2 | XP_005247543.1 | 3 | 420 | |||
XM_005247488.2 | XP_005247545.1 | 3 | 525 | |||
XM_017006452.1 | XP_016861941.1 | 5 | 1309 | |||
XM_017006453.1 | XP_016861942.1 | 3 | 575 | |||
XM_017006454.1 | XP_016861943.1 | 5 | 1298 | |||
XM_017006455.1 | XP_016861944.1 | 4 | 933 | |||
XM_017006456.1 | XP_016861945.1 | 5 | 659 | |||
XM_017006457.1 | XP_016861946.1 | 4 | 607 | |||
XM_017006458.1 | XP_016861947.1 | 5 | 1299 | |||
XM_017006459.1 | XP_016861948.1 | 4 | 918 | |||
GenBank mRNA | AF063591.1 | - | 4 | 524 | ||
AK074972.1 | - | 4 | 498 | |||
AK129797.1 | - | 3 | 415 | |||
AK293399.1 | - | 3 | 420 | |||
AK297194.1 | - | 5 | 599 | |||
AK312563.1 | - | 4 | 609 | |||
AL832529.1 | - | 4 | 518 | |||
AY603771.1 | - | 3 | 467 | |||
BC022522.1 | - | 4 | 526 | 4794050 | ||
BC031103.1 | - | 4 | 502 | 5301702 | ||
BI547787.1 | - | 5 | 604 |