Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015052.2 | NP_001015052.1 | 4 | 811 | ||
NM_001015054.2 | NP_001015054.1 | 4 | 754 | |||
NM_002434.3 | NP_002425.2 | 5 | 1004 | |||
GenBank mRNA | AW083039.1 | - | 1 | 298 | ||
AY258284.1 | - | 4 | 720 | |||
BC014991.1 | - | 4 | 750 | 4909112 | ||
L10752.1 | - | 4 | 672 | |||
M71215.1 | - | 3 | 483 | |||
M74905.1 | - | 4 | 832 | |||
S51033.1 | - | 3 | 605 | |||
X56528.1 | - | 3 | 483 |