Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166460.1 | NP_001159932.1 | 10 | 1214 | ||
NM_001166461.1 | NP_001159933.1 | 10 | 1205 | |||
NM_001166462.1 | NP_001159934.1 | 11 | 1414 | |||
NM_002436.3 | NP_002427.1 | 10 | 1265 | |||
XM_011531167.1 | XP_011529469.1 | 9 | 1178 | |||
XM_011531169.1 | XP_011529471.1 | 6 | 837 | |||
GenBank mRNA | AK093702.1 | - | 1 | 1597 | ||
AK223396.1 | - | 10 | 1245 | |||
AK290246.1 | - | 10 | 1160 | |||
AK303111.1 | - | 10 | 1080 | |||
AK304538.1 | - | 10 | 1172 | |||
AK311541.1 | - | 11 | 1145 | |||
AK312296.1 | - | 10 | 1136 | |||
AK315957.1 | - | 11 | 1263 | |||
AY423731.1 | - | 10 | 1137 | |||
AY634686.1 | - | 10 | 1075 | |||
BC002392.2 | - | 10 | 1141 | 2820598 | ||
M64925.1 | - | 10 | 1214 |