Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166460.1 | NP_001159932.1 | 9 | 997 | ||
NM_001166461.1 | NP_001159933.1 | 9 | 988 | |||
NM_001166462.1 | NP_001159934.1 | 10 | 1197 | |||
NM_002436.3 | NP_002427.1 | 9 | 1048 | |||
XM_011531167.1 | XP_011529469.1 | 8 | 961 | |||
XM_011531169.1 | XP_011529471.1 | 5 | 620 | |||
GenBank mRNA | AK223396.1 | - | 9 | 1028 | ||
AK290246.1 | - | 9 | 943 | |||
AK303111.1 | - | 9 | 863 | |||
AK304538.1 | - | 9 | 955 | |||
AK311541.1 | - | 10 | 928 | |||
AK312296.1 | - | 9 | 919 | |||
AK315957.1 | - | 10 | 1046 | |||
AY423731.1 | - | 9 | 920 | |||
AY634686.1 | - | 9 | 858 | |||
BC002392.2 | - | 9 | 924 | 2820598 | ||
M64925.1 | - | 9 | 997 |