Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002444.2 | NP_002435.1 | 12 | 1589 | ||
XM_005262269.2 | XP_005262326.1 | 12 | 1610 | |||
XM_011530959.1 | XP_011529261.1 | 12 | 1505 | |||
XM_017029545.1 | XP_016885034.1 | 14 | 1733 | |||
XM_017029546.1 | XP_016885035.1 | 13 | 1629 | |||
GenBank mRNA | AK297165.1 | - | 5 | 370 | ||
AK309986.1 | - | 2 | 122 | |||
BC017293.2 | - | 12 | 1542 | 4908580 | ||
EU794636.1 | - | 12 | 1542 | |||
M69066.1 | - | 12 | 1491 |