Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK309266.1 | - | 5 | 408 | ||
AK312361.1 | - | 6 | 500 | |||
BC009806.2 | - | 6 | 461 | 3508998 | ||
BC050420.1 | - | 6 | 486 | 5788176 | ||
BQ959308.1 | - | 6 | 607 | |||
J04031.1 | - | 6 | 461 |