Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005369.1 | NP_001005369.1 | 15, 16 | 2607 | ||
NM_001321001.1 | NP_001307930.1 | 14, 15 | 2142 | |||
NM_001321002.1 | NP_001307931.1 | 14, 15 | 2114 | |||
NM_001321003.1 | NP_001307932.1 | 15, 16 | 2324 | |||
NM_001321004.1 | NP_001307933.1 | 13, 14 | 2376 | |||
NM_001321005.1 | NP_001307934.1 | 13, 14 | 2025 | |||
NM_002453.2 | NP_002444.2 | 14, 15 | 2186 | |||
XM_005264335.3 | XP_005264392.1 | 12, 13 | 1985 | |||
XM_011532871.2 | XP_011531173.1 | 13, 14 | 2019 | |||
XM_017004162.1 | XP_016859651.1 | 15, 16 | 2315 | |||
XM_017004163.1 | XP_016859652.1 | 14, 15 | 2391 | |||
XM_017004164.1 | XP_016859653.1 | 13, 14 | 2003 | |||
XM_017004165.1 | XP_016859654.1 | 14, 15 | 2218 | |||
XM_017004166.1 | XP_016859655.1 | 14, 15 | 2410 | |||
XM_017004167.1 | XP_016859656.1 | 10, 11 | 2487 | |||
GenBank mRNA | AF494407.1 | - | 14, 15 | 2166 | ||
AK307993.1 | - | 14, 15 | 2166 | |||
BU681583.1 | - | 2, 3 | 355 | |||
BU684812.1 | - | 2, 3 | 355 | |||
L34600.1 | - | 14, 15 | 2140 |