Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000254.2 | NP_000245.2 | 13 | 1530 | ||
NM_001291939.1 | NP_001278868.1 | 13 | 1530 | |||
XM_005273141.4 | XP_005273198.1 | 13 | 1499 | |||
XM_006711770.2 | XP_006711833.1 | 7 | 478 | |||
XM_011544194.2 | XP_011542496.1 | 12 | 2292 | |||
XM_017001329.1 | XP_016856818.1 | 12 | 2292 | |||
XM_017001330.1 | XP_016856819.1 | 12 | 2292 | |||
GenBank mRNA | AK299904.1 | - | 13 | 1530 | ||
AK307390.1 | - | 13 | 1519 | |||
BC130616.1 | - | 13 | 1129 | 40146647 | ||
BC136440.1 | - | 13 | 1129 | 9020429 | ||
BC144095.1 | - | 13 | 1129 | 9052615 | ||
BC144097.1 | - | 13 | 1129 | 9052617 | ||
U71285.1 | - | 13 | 1170 | |||
U73338.1 | - | 13 | 1501 | |||
U75743.1 | - | 13 | 1393 |