Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005592.3 | NP_005583.1 | 9, 10 | 1325 | ||
XM_005251994.3 | XP_005252051.1 | 10, 11 | 1937 | |||
XM_005251995.3 | XP_005252052.1 | 10, 11 | 1936 | |||
XM_005251996.3 | XP_005252053.1 | 9, 10 | 1905 | |||
XM_011518708.2 | XP_011517010.1 | 1, 2 | 70 | |||
GenBank mRNA | AF006464.1 | - | 9, 10 | 1237 |