Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002472.2 | NP_002463.2 | 40 | 5858 | ||
GenBank mRNA | M36769.1 | - | 39 | 5836 | ||
X51592.1 | - | 26 | 4263 | |||
Z38133.1 | - | 39 | 5836 |