Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000432.3 | NP_000423.2 | 6, 7 | 473 | ||
GenBank mRNA | AF020768.1 | - | 5, 6 | 407 | ||
AK312013.1 | - | 6, 7 | 450 | |||
BC015821.1 | - | 6, 7 | 455 | 4245364 | ||
BC031006.1 | - | 6, 7 | 450 | 4292208 | ||
BC031008.1 | - | 6, 7 | 452 | 4297454 | ||
CR456962.1 | - | 5, 6 | 402 | |||
CR541957.1 | - | 5, 6 | 402 | |||
M22815.1 | - | 7, 8 | 421 | |||
S69022.1 | - | 6, 7 | 479 | |||
X14332.1 | - | 6, 7 | 399 | |||
X57542.1 | - | 7, 8 | 421 | |||
X66141.1 | - | 6, 7 | 432 |