Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000260.3 | NP_000251.3 | 17 | 2300 | ||
NM_001127179.2 | NP_001120651.2 | 17 | 2300 | |||
NM_001127180.1 | NP_001120652.1 | 17 | 2294 | |||
XM_011545044.2 | XP_011543346.1 | 17 | 2372 | |||
XM_011545046.2 | XP_011543348.2 | 16 | 2133 | |||
XM_011545050.2 | XP_011543352.1 | 14 | 1987 | |||
XM_017017778.1 | XP_016873267.1 | 16 | 2133 | |||
XM_017017779.1 | XP_016873268.1 | 16 | 2133 | |||
XM_017017780.1 | XP_016873269.1 | 16 | 2133 | |||
XM_017017781.1 | XP_016873270.1 | 16 | 2133 | |||
XM_017017782.1 | XP_016873271.1 | 16 | 2133 | |||
XM_017017783.1 | XP_016873272.1 | 16 | 2133 | |||
XM_017017784.1 | XP_016873273.1 | 16 | 2133 | |||
XM_017017785.1 | XP_016873274.1 | 14 | 1902 | |||
XM_017017786.1 | XP_016873275.1 | 16 | 2133 | |||
XM_017017787.1 | XP_016873276.1 | 16 | 2133 | |||
XM_017017788.1 | XP_016873277.1 | 16 | 2133 | |||
GenBank mRNA | BC023212.1 | - | 2 | 308 | ||
BC144519.1 | - | 18 | 2342 | |||
U34227.1 | - | 17 | 2295 | |||
U39226.1 | - | 17 | 2300 | |||
U55208.1 | - | 17 | 2295 | |||
U55209.1 | - | 17 | 2295 |