Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004539.3 | NP_004530.1 | 12 | 1508 | ||
XM_005266700.1 | XP_005266757.1 | 12 | 1286 | |||
GenBank mRNA | AJ000334.1 | - | 12 | 1353 | ||
AK129897.1 | - | 12 | 1310 | |||
AK222835.1 | - | 12 | 1355 | |||
AK297777.1 | - | 10 | 939 | |||
BC001687.1 | - | 12 | 1357 | 2987840 | ||
D84273.1 | - | 12 | 1312 |