Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004539.3 | NP_004530.1 | 6 | 679 | ||
XM_005266700.1 | XP_005266757.1 | 6 | 457 | |||
GenBank mRNA | AJ000334.1 | - | 6 | 524 | ||
AK129897.1 | - | 6 | 482 | |||
AK222835.1 | - | 6 | 526 | |||
AK293985.1 | - | 5 | 459 | |||
AK294763.1 | - | 6 | 510 | |||
BC001687.1 | - | 6 | 528 | 2987840 | ||
D84273.1 | - | 6 | 483 |