Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004539.3 | NP_004530.1 | 8 | 827 | ||
XM_005266700.1 | XP_005266757.1 | 8 | 605 | |||
GenBank mRNA | AJ000334.1 | - | 8 | 672 | ||
AK222835.1 | - | 8 | 674 | |||
AK294364.1 | - | 7 | 603 | |||
AK294763.1 | - | 8 | 658 | |||
BC001687.1 | - | 8 | 676 | 2987840 | ||
D84273.1 | - | 8 | 631 |