Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000615.6 | NP_000606.3 | 15 | 2268 | ||
NM_001076682.3 | NP_001070150.1 | 18 | 2373 | |||
NM_001242607.1 | NP_001229536.1 | 17 | 2376 | |||
NM_001242608.1 | NP_001229537.1 | 15 | 2268 | |||
NM_181351.4 | NP_851996.2 | 16 | 2298 | |||
GenBank mRNA | AB209443.1 | - | 15 | 2147 | ||
AK057509.1 | - | 15 | 2371 | |||
AK292453.1 | - | 17 | 1933 | |||
AK299865.1 | - | 6 | 479 | |||
AK303232.1 | - | 9 | 639 | |||
AK314589.1 | - | 17 | 2348 | |||
BC014205.2 | - | 10 | 1162 | |||
BC047244.1 | - | 16 | 2110 | 5590188 | ||
M17409.1 | - | 8 | 537 | |||
M17410.1 | - | 4 | 429 | |||
M22094.1 | - | 9 | 776 | |||
S71824.1 | - | 15 | 2103 | |||
U63041.1 | - | 15 | 1939 | |||
X16841.1 | - | 18 | 2152 |