Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002487.2 | NP_002478.1 | 1 | 475 | ||
GenBank mRNA | AK129654.1 | - | 2 | 373 | ||
AK312779.1 | - | 1 | 469 | |||
BC008750.1 | - | 1 | 452 | 3347128 | ||
KJ534888.1 | - | 1 | 389 | |||
KU178221.1 | - | 1 | 389 | |||
KU178222.1 | - | 2 | 191 | |||
U35139.1 | - | 1 | 447 |