Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002487.2 | NP_002478.1 | 1 | 894 | ||
GenBank mRNA | AK129654.1 | - | 2 | 792 | ||
AK312779.1 | - | 1 | 888 | |||
BC008750.1 | - | 1 | 871 | 3347128 | ||
KJ534888.1 | - | 1 | 808 | |||
KJ535010.1 | - | 2 | 408 | |||
KU178221.1 | - | 1 | 808 | |||
KU178222.1 | - | 2 | 610 | |||
KU178223.1 | - | 2 | 486 | |||
KU178224.1 | - | 2 | 392 | |||
KU178225.1 | - | 2 | 325 | |||
U35139.1 | - | 1 | 866 |