Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199958.1 | NP_001186887.1 | 2 | 165 | ||
NM_002492.3 | NP_002483.1 | 2 | 165 | |||
GenBank mRNA | AF047181.1 | - | 2 | 142 | ||
AK313822.1 | - | 2 | 143 | |||
AW955118.1 | - | 2 | 122 | |||
BC005271.1 | - | 2 | 156 | 3997377 | ||
BC009796.1 | - | 2 | 160 | 4284776 | ||
BC017909.1 | - | 2 | 155 | |||
BG776403.1 | - | 2 | 171 | |||
DA112751.1 | - | 2 | 168 |