Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166102.1 | NP_001159574.1 | 8 | 1234 | ||
NM_007103.3 | NP_009034.2 | 8 | 1261 | |||
GenBank mRNA | AF053070.1 | - | 8 | 1177 | ||
AF092131.1 | - | 8 | 1187 | |||
AK055875.1 | - | 5 | 2340 | |||
AK223061.1 | - | 8 | 1155 | |||
AK293522.1 | - | 7 | 1072 | |||
AK300726.1 | - | 6 | 768 | |||
AK316122.1 | - | 8 | 1102 | |||
BC007619.1 | - | 5 | 778 | |||
BC008146.1 | - | 8 | 1146 | 4156629 | ||
BC015645.2 | - | 8 | 1141 | 4648781 | ||
CR456739.1 | - | 8 | 1108 | |||
DB532599.1 | - | 2 | 57 | |||
S67973.1 | - | 6 | 730 |