Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002495.3 | NP_002486.1 | 3, 4 | 391 | ||
NR_134473.1 | 4, 5 | 563 | ||||
NR_134474.1 | 4, 5 | 480 | ||||
NR_134475.1 | 4, 5 | 515 | ||||
GenBank mRNA | AF020351.1 | - | 3, 4 | 369 | ||
AI001139.1 | - | 2 | 284 | |||
AK307973.1 | - | 3, 4 | 369 | |||
BC005270.1 | - | 3, 4 | 385 | 3681312 | ||
BU954271.1 | - | 3, 4 | 383 | |||
CD692133.1 | - | 4, 5 | 531 | |||
DA609203.1 | - | 4, 5 | 480 | |||
DB489661.1 | - | 3, 4 | 390 | |||
KU178226.1 | - | 3, 4 | 361 | |||
KU178227.1 | - | 4, 5 | 533 |