Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002495.3 | NP_002486.1 | 4 | 420 | ||
NR_134473.1 | 5 | 592 | ||||
NR_134474.1 | 5 | 509 | ||||
NR_134475.1 | 5 | 544 | ||||
GenBank mRNA | AF020351.1 | - | 4 | 398 | ||
AI001139.1 | - | 2 | 255 | |||
BC005270.1 | - | 4 | 414 | 3681312 | ||
BU954271.1 | - | 4 | 412 | |||
CD692133.1 | - | 5 | 560 | |||
DA609203.1 | - | 5 | 509 | |||
DB489661.1 | - | 4 | 419 | |||
KU178226.1 | - | 4 | 390 | |||
KU178227.1 | - | 5 | 562 |