Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002496.3 | NP_002487.1 | 6 | 548 | ||
XM_005274013.1 | XP_005274070.1 | 6 | 645 | |||
XM_005274014.2 | XP_005274071.1 | 6 | 672 | |||
XM_011545053.2 | XP_011543355.1 | 6 | 583 | |||
GenBank mRNA | AK223114.1 | - | 6 | 538 | ||
AK314546.1 | - | 6 | 473 | |||
BC119753.1 | - | 2 | 105 | |||
BC119754.2 | - | 6 | 477 | 40119290 | ||
U65579.1 | - | 6 | 471 |