Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002496.3 | NP_002487.1 | 7 | 632 | ||
XM_005274013.1 | XP_005274070.1 | 7 | 729 | |||
XM_005274014.2 | XP_005274071.1 | 7 | 756 | |||
XM_011545053.2 | XP_011543355.1 | 7 | 667 | |||
GenBank mRNA | AK223114.1 | - | 7 | 622 | ||
AK314546.1 | - | 7 | 557 | |||
BC119753.1 | - | 3 | 189 | |||
BC119754.2 | - | 7 | 561 | 40119290 | ||
U65579.1 | - | 7 | 555 |