Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284338.1 | NP_001271267.1 | 19 | 3722 | ||
NM_001284339.1 | NP_001271268.1 | 19 | 3674 | |||
NM_001284340.1 | NP_001271269.1 | 18 | 3671 | |||
NM_006154.3 | NP_006145.2 | 23 | 2356 | |||
NM_198400.3 | NP_940682.2 | 16 | 3506 | |||
NR_104302.1 | 17 | 3609 | ||||
XM_011521624.2 | XP_011519926.1 | 22 | 2121 | |||
XM_011521625.2 | XP_011519927.1 | 24 | 2294 | |||
XM_011521626.1 | XP_011519928.1 | 22 | 2138 | |||
XM_011521627.1 | XP_011519929.1 | 20 | 1961 | |||
GenBank mRNA | AK304394.1 | - | 23 | 2222 | ||
AK307541.1 | - | 19 | 1973 | |||
AL832063.1 | - | 16 | 3507 | |||
AL832359.1 | - | 18 | 2194 | |||
AL833293.1 | - | 18 | 2947 | |||
AL833300.1 | - | 19 | 2704 | |||
AY550969.1 | - | 23 | 2235 | |||
BC035707.1 | - | 23 | 2200 | |||
BC136605.1 | - | 19 | 3699 | 9020595 | ||
BC144284.1 | - | 19 | 3699 | 9052804 | ||
BC144285.1 | - | 19 | 3651 | 9052805 | ||
BC152452.1 | - | 23 | 2247 | 8862584 | ||
BC152562.1 | - | 23 | 2247 | 8860509 | ||
D42055.1 | - | 23 | 2247 |