Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199397.1 | NP_001186326.1 | 35 | 4374 | ||
NM_001199398.1 | NP_001186327.1 | 34 | 4242 | |||
NM_001199399.1 | NP_001186328.1 | 32 | 4083 | |||
NM_001199400.1 | NP_001186329.1 | 32 | 4236 | |||
NM_012224.2 | NP_036356.1 | 33 | 4368 | |||
XM_006714228.1 | XP_006714291.1 | 34 | 4199 | |||
XM_011532003.1 | XP_011530305.1 | 34 | 4292 | |||
XM_011532004.1 | XP_011530306.1 | 33 | 4160 | |||
XM_017008249.1 | XP_016863738.1 | 34 | 4278 | |||
XM_017008250.1 | XP_016863739.1 | 33 | 4396 | |||
XM_017008251.1 | XP_016863740.1 | 33 | 4194 | |||
XM_017008252.1 | XP_016863741.1 | 31 | 4214 | |||
XM_017008253.1 | XP_016863742.1 | 34 | 4224 | |||
XM_017008254.1 | XP_016863743.1 | 27 | 3334 | |||
GenBank mRNA | AB067488.1 | - | 33 | 4204 | ||
AF155113.1 | - | 17 | 2252 | |||
AK025658.1 | - | 8 | 1122 | |||
AL050385.1 | - | 34 | 4198 | |||
BC037790.2 | - | 7 | 1034 | |||
BC114491.1 | - | 32 | 3843 | 40082305 | ||
CD358162.1 | - | 3 | 373 | |||
CR933642.1 | - | 35 | 4332 | |||
CR936672.1 | - | 32 | 4194 |