Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002500.4 | NP_002491.2 | 2 | 452 | ||
GenBank mRNA | AB593068.1 | - | 2 | 304 | ||
AB593069.1 | - | 2 | 306 | |||
AB593070.1 | - | 2 | 304 | |||
AB593071.1 | - | 2 | 304 | |||
AK313799.1 | - | 2 | 304 | |||
BC009046.1 | - | 2 | 273 | 3873419 | ||
BT019731.1 | - | 1 | 205 | |||
D82347.1 | - | 2 | 314 | |||
DA486317.1 | - | 2 | 442 | |||
DB472071.1 | - | 2 | 397 | |||
KU178230.1 | - | 1 | 205 |