Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEUROD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002500.4 | NP_002491.2 | 2 | 817 | ||
GenBank mRNA | AB593068.1 | - | 2 | 669 | ||
AB593069.1 | - | 2 | 671 | |||
AB593070.1 | - | 2 | 669 | |||
AB593071.1 | - | 2 | 669 | |||
AK313799.1 | - | 2 | 669 | |||
BC009046.1 | - | 2 | 638 | 3873419 | ||
BT019731.1 | - | 1 | 570 | |||
D82347.1 | - | 2 | 679 | |||
KU178230.1 | - | 1 | 570 | |||
U36472.1 | - | 1 | 134 | |||
U80578.1 | - | 1 | 309 |