Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004555.3 | NP_004546.1 | 9 | 3380 | ||
NM_173163.2 | NP_775186.1 | 9 | 3380 | |||
NM_173165.2 | NP_775188.1 | 9 | 3380 | |||
GenBank mRNA | AB209824.1 | - | 7 | 2007 | ||
AK290370.1 | - | 9 | 3120 | |||
BC001050.2 | - | 9 | 3098 | 3505967 | ||
EU887605.1 | - | 9 | 3064 | |||
EU887606.1 | - | 9 | 3064 | |||
EU887607.1 | - | 9 | 3064 | |||
EU887608.1 | - | 9 | 3391 | |||
EU887609.1 | - | 9 | 3391 | |||
EU887610.1 | - | 9 | 3391 | |||
EU887611.1 | - | 9 | 3109 | |||
EU887612.1 | - | 9 | 3109 | |||
EU887613.1 | - | 9 | 3109 | |||
EU887614.1 | - | 9 | 3017 | |||
EU887615.1 | - | 9 | 3017 | |||
EU887616.1 | - | 9 | 3017 | |||
EU887617.1 | - | 9 | 3022 | |||
EU887618.1 | - | 9 | 3022 | |||
EU887619.1 | - | 9 | 3022 | |||
EU887620.1 | - | 9 | 3059 | |||
EU887621.1 | - | 9 | 3059 | |||
EU887622.1 | - | 9 | 3059 | |||
EU887623.1 | - | 9 | 3463 | |||
EU887624.1 | - | 9 | 3463 | |||
EU887625.1 | - | 9 | 3463 | |||
L41067.1 | - | 9 | 3274 | |||
U14510.1 | - | 9 | 3088 | |||
U85428.1 | - | 9 | 3088 | |||
U85429.1 | - | 9 | 3088 | |||
U85430.1 | - | 9 | 3088 |