Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077494.3 | NP_001070962.1 | 3 | 328 | ||
NM_001261403.2 | NP_001248332.1 | 2 | 423 | |||
NM_001288724.1 | NP_001275653.1 | 3 | 622 | |||
NM_001322934.1 | NP_001309863.1 | 3 | 332 | |||
NM_001322935.1 | NP_001309864.1 | 3 | 622 | |||
NM_002502.5 | NP_002493.3 | 3 | 332 | |||
XM_017016278.1 | XP_016871767.1 | 3 | 617 | |||
GenBank mRNA | AK292654.1 | - | 2 | 276 | ||
BC002844.2 | - | 3 | 159 | 3634575 | ||
BT009769.1 | - | 2 | 59 | |||
BY799958.2 | - | 3 | 260 | |||
DA349980.1 | - | 2 | 344 | |||
DA408461.1 | - | 3 | 259 | |||
DA711063.1 | - | 3 | 326 | |||
S76638.1 | - | 3 | 309 | |||
U09609.2 | - | 2 | 59 | |||
X61498.1 | - | 3 | 222 | |||
X61499.1 | - | 3 | 222 |