Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFYC SIRNA Assays ›
Gene Symbol:
NFYC
Gene Name:
nuclear transcription factor Y subunit gamma
Gene Aliases:
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
Chromosome Location:
Chr.1: 40691570 - 40771603 on Build GRCh38
UniGene ID:
Hs.713051
Species:
Human
siRNA ID
HSS107175
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFYC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142587.1
NP_001136059.1
5
655
NM_001142588.1
NP_001136060.1
5
655
NM_001142589.1
NP_001136061.1
4
541
NM_001142590.1
NP_001136062.1
10
1032
NM_001308114.1
NP_001295043.1
4
357
NM_001308115.1
NP_001295044.1
5
651
NM_014223.4
NP_055038.2
5
655
XM_005270894.1
XP_005270951.1
5
651
XM_005270895.3
XP_005270952.3
5
507
XM_006710658.2
XP_006710721.2
5
507
XM_006710660.2
XP_006710723.2
5
507
XM_006710661.1
XP_006710724.1
5
536
XM_006710662.2
XP_006710725.2
5
507
XM_011541516.1
XP_011539818.1
5
456
XM_011541517.1
XP_011539819.1
5
507
XM_017001364.1
XP_016856853.1
5
676
XM_017001365.1
XP_016856854.1
5
536
XM_017001366.1
XP_016856855.1
5
610
XM_017001367.1
XP_016856856.1
5
651
XM_017001368.1
XP_016856857.1
5
651
XM_017001369.1
XP_016856858.1
6
1008
GenBank mRNA
AB208975.1
-
5
482
AF191744.1
-
4
356
AK000346.1
-
5
608
AK127677.1
-
10
1027
AK300774.1
-
5
458
AK301385.1
-
4
425
BC005003.1
-
5
504
3502903
BT020081.1
-
4
349
D85425.1
-
5
432
D89986.1
-
5
417
U62296.1
-
4
349
U78774.1
-
6
523
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein folding
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
protein binding
transcription factor binding
transcription regulatory region DNA binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline