Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002507.3 | NP_002498.1 | 4 | 929 | ||
GenBank mRNA | AK303278.1 | - | 4 | 997 | ||
AK313654.1 | - | 4 | 929 | |||
M14764.1 | - | 4 | 917 |